Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCCTTTGAAGGAGAGGGCCCTG[C/G]AGCCCTCTCCTTAGCAATTAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 150330 | ||||||||||||||||||||
Literature Links: |
LMNA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMNA - lamin A/C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257374.2 | Intron | NP_001244303.1 | ||||
NM_001282624.1 | Intron | NP_001269553.1 | ||||
NM_001282625.1 | Intron | NP_001269554.1 | ||||
NM_001282626.1 | Intron | NP_001269555.1 | ||||
NM_005572.3 | Intron | NP_005563.1 | ||||
NM_170707.3 | Intron | NP_733821.1 | ||||
NM_170708.3 | Intron | NP_733822.1 | ||||
XM_011509533.1 | Intron | XP_011507835.1 | ||||
XM_011509534.1 | Intron | XP_011507836.1 |