Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAATCACAGAGCAGCAGCCCCCA[A/G]ACTTGGCAGGAAGCATGGCAGCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607079 | ||||||||||||||||||||
Literature Links: |
RHBG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHBG - Rh family B glycoprotein (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256395.1 | Intron | NP_001243324.1 | ||||
NM_001256396.1 | Intron | NP_001243325.1 | ||||
NM_020407.4 | Intron | NP_065140.3 | ||||
XM_011509799.2 | Intron | XP_011508101.1 | ||||
XM_011509800.2 | Intron | XP_011508102.1 | ||||
XM_017001857.1 | Intron | XP_016857346.1 | ||||
XM_017001858.1 | Intron | XP_016857347.1 | ||||
XM_017001859.1 | Intron | XP_016857348.1 |