Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGCCCCGCTGGCTCCCGCCTGC[G/T]CCTGCTCCTGCTCCTGCTGCTGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603181 MIM: 108960 | ||||||||||||||||||||
Literature Links: |
ILF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ILF2 - interleukin enhancer binding factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPR1 - natriuretic peptide receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000906.3 | 530 | Missense Mutation | CGC,CTC | R,L 13 | NP_000897.3 | |
XM_005245218.1 | 530 | Missense Mutation | CGC,CTC | R,L 13 | XP_005245275.1 | |
XM_017001374.1 | 530 | Missense Mutation | CGC,CTC | R,L 13 | XP_016856863.1 |