Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTGTTGGGATTACAGGCTTGGT[C/T]CACCATGCCCCACCTGTAGGTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP42 - ubiquitin specific peptidase 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032172.2 | Intron | NP_115548.1 | ||||
XM_005249883.4 | Intron | XP_005249940.1 | ||||
XM_006715790.1 | Intron | XP_006715853.1 | ||||
XM_006715791.2 | Intron | XP_006715854.1 | ||||
XM_011515573.1 | Intron | XP_011513875.1 | ||||
XM_011515574.1 | Intron | XP_011513876.1 | ||||
XM_011515577.1 | Intron | XP_011513879.1 | ||||
XM_011515578.1 | Intron | XP_011513880.1 |