Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194540 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIVEP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIVEP1 - human immunodeficiency virus type I enhancer binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002114.3 | Intron | NP_002105.3 | ||||
XM_011514546.2 | Intron | XP_011512848.1 | ||||
XM_011514547.2 | Intron | XP_011512849.1 | ||||
XM_011514548.2 | Intron | XP_011512850.1 | ||||
XM_011514549.2 | Intron | XP_011512851.1 | ||||
XM_011514550.2 | Intron | XP_011512852.1 | ||||
XM_011514551.2 | Intron | XP_011512853.1 | ||||
XM_011514552.2 | Intron | XP_011512854.1 | ||||
XM_011514553.1 | Intron | XP_011512855.1 | ||||
XM_011514555.2 | Intron | XP_011512857.1 | ||||
XM_017010800.1 | Intron | XP_016866289.1 | ||||
XM_017010801.1 | Intron | XP_016866290.1 | ||||
XM_017010802.1 | Intron | XP_016866291.1 | ||||
XM_017010803.1 | Intron | XP_016866292.1 | ||||
XM_017010804.1 | Intron | XP_016866293.1 |