Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCTGATTATGTCCATTGAACCA[C/T]ATACTAAAGATTTAATGTAGTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615734 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR47 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
WDR47 - WD repeat domain 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142550.1 | Intron | NP_001136022.1 | ||||
NM_001142551.1 | Intron | NP_001136023.1 | ||||
NM_014969.5 | Intron | NP_055784.3 | ||||
XM_006710460.3 | Intron | XP_006710523.1 | ||||
XM_011541027.2 | Intron | XP_011539329.1 | ||||
XM_011541028.2 | Intron | XP_011539330.1 | ||||
XM_011541029.1 | Intron | XP_011539331.1 | ||||
XM_011541030.1 | Intron | XP_011539332.1 | ||||
XM_011541031.1 | Intron | XP_011539333.1 | ||||
XM_017000696.1 | Intron | XP_016856185.1 | ||||
XM_017000697.1 | Intron | XP_016856186.1 |