Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCCCTTTCGGGGCGGAAGACTA[C/T]GTTTGAGCATCTCACTGAGGTGCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604345 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN1A2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAN1A2 - mannosidase alpha class 1A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006699.3 | 368 | UTR 5 | NP_006690.1 | |||
XM_006710302.2 | 368 | UTR 5 | XP_006710365.1 | |||
XM_011540536.2 | 368 | UTR 5 | XP_011538838.1 | |||
XM_017000115.1 | 368 | UTR 5 | XP_016855604.1 |
Set Membership: |
HapMap |