Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAAGGGGAGGTTGAGGGCCTGA[G/T]AAGTTGAGTGTGGGAAGGCATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611764 MIM: 604267 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNFN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNFN - cornifelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032488.3 | Intron | NP_115877.2 | ||||
XM_005259332.2 | Intron | XP_005259389.1 | ||||
XM_011527396.1 | Intron | XP_011525698.1 | ||||
XM_011527397.1 | Intron | XP_011525699.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101930071 - uncharacterized LOC101930071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEGF8 - multiple EGF like domains 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |