Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAE1 - SUMO1 activating enzyme subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145713.1 | Intron | NP_001139185.1 | ||||
NM_001145714.1 | Intron | NP_001139186.1 | ||||
NM_005500.2 | Intron | NP_005491.1 | ||||
XM_006722963.2 | Intron | XP_006723026.1 | ||||
XM_006722965.3 | Intron | XP_006723028.1 | ||||
XM_017026135.1 | Intron | XP_016881624.1 | ||||
XM_017026136.1 | Intron | XP_016881625.1 |