Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCTTCTTTTTTAACCTTTAAAT[G/T]AGATGGAATAATGTTGTTTGCTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611036 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C12orf40 - chromosome 12 open reading frame 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031748.3 | 6544 | Intron | NP_001026918.2 | |||
NM_001319247.1 | 6544 | Intron | NP_001306176.1 | |||
XM_005268806.3 | 6544 | Intron | XP_005268863.1 | |||
XM_011538231.2 | 6544 | Intron | XP_011536533.1 |
SLC2A13 - solute carrier family 2 member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052885.3 | 6544 | UTR 3 | NP_443117.3 | |||
XM_011537847.2 | 6544 | UTR 3 | XP_011536149.1 | |||
XM_011537849.2 | 6544 | Intron | XP_011536151.1 | |||
XM_011537850.2 | 6544 | Intron | XP_011536152.1 | |||
XM_017018764.1 | 6544 | Intron | XP_016874253.1 | |||
XM_017018765.1 | 6544 | Intron | XP_016874254.1 | |||
XM_017018766.1 | 6544 | Intron | XP_016874255.1 |
Set Membership: |
HapMap |