Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGCCAAGACCCACTGACATTA[C/G]CCGAATATGATTATTTCTCTTACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611681 | ||||||||||||||||||||
Literature Links: |
ADAMTS20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS20 - ADAM metallopeptidase with thrombospondin type 1 motif 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025003.3 | Intron | NP_079279.3 | ||||
XM_011538754.2 | Intron | XP_011537056.1 | ||||
XM_017019979.1 | Intron | XP_016875468.1 |