Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAGTATTAGAACTATAAAAGAA[A/G]CACCATCTAATTAATTCTATCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIGD1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIGD1C - HIG1 hypoxia inducible domain family member 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109619.1 | Intron | NP_001103089.1 | ||||
XM_011538649.2 | Intron | XP_011536951.1 | ||||
XM_017019783.1 | Intron | XP_016875272.1 | ||||
XM_017019784.1 | Intron | XP_016875273.1 | ||||
XM_017019785.1 | Intron | XP_016875274.1 |
METTL7A - methyltransferase like 7A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |