Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTGAACACAGGTTATAATGGGT[A/G]GCACTGGTGGTATGAGAGGGTGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFCP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TFCP2 - transcription factor CP2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173452.1 | 3018 | UTR 3 | NP_001166923.1 | |||
NM_001173453.1 | 3018 | UTR 3 | NP_001166924.1 | |||
NM_005653.4 | 3018 | UTR 3 | NP_005644.2 |
Set Membership: |
HapMap |