Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGGCCCATTATCCTTTAATGTT[C/G]CATTACATATCTTCCCAAAACAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600469 MIM: 611153 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCBP1 - nuclear cap binding protein subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002486.4 | Intron | NP_002477.1 | ||||
XM_011518715.2 | Intron | XP_011517017.1 | ||||
XM_017014735.1 | Intron | XP_016870224.1 | ||||
XM_017014736.1 | Intron | XP_016870225.1 | ||||
XM_017014737.1 | Intron | XP_016870226.1 | ||||
XM_017014738.1 | Intron | XP_016870227.1 |
XPA - XPA, DNA damage recognition and repair factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000380.3 | Intron | NP_000371.1 | ||||
XM_006717278.1 | Intron | XP_006717341.1 | ||||
XM_017015114.1 | Intron | XP_016870603.1 |