Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCACTAGATGGCGCTGTGAGG[C/T]GCAGTGGGAAGAGTAGCTGGAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRMO PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRMO - tRNA methyltransferase O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016481.4 | Intron | NP_057565.3 | ||||
XM_005252047.3 | Intron | XP_005252104.1 | ||||
XM_005252048.4 | Intron | XP_005252105.1 | ||||
XM_005252049.4 | Intron | XP_005252106.1 | ||||
XM_005252050.4 | Intron | XP_005252107.1 | ||||
XM_011518776.2 | Intron | XP_011517078.1 | ||||
XM_011518777.1 | Intron | XP_011517079.1 |