Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCGCATCTCTTCTTGCTGGGCCA[A/G]TTCCCACAGGCTCTTCCAGCTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607340 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABBR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABBR2 - gamma-aminobutyric acid type B receptor subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005458.7 | Intron | NP_005449.5 | ||||
XM_005252316.4 | Intron | XP_005252373.1 | ||||
XM_017015331.1 | Intron | XP_016870820.1 | ||||
XM_017015332.1 | Intron | XP_016870821.1 |
Set Membership: |
HapMap |