Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGGTTAGGTCTGGGCAGGGTC[C/G]TTCACTTCCCTCTACTATCACCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610786 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRCIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SRCIN1 - SRC kinase signaling inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025248.2 | Intron | NP_079524.2 | ||||
XM_017025169.1 | Intron | XP_016880658.1 | ||||
XM_017025170.1 | Intron | XP_016880659.1 | ||||
XM_017025171.1 | Intron | XP_016880660.1 | ||||
XM_017025172.1 | Intron | XP_016880661.1 | ||||
XM_017025173.1 | Intron | XP_016880662.1 | ||||
XM_017025174.1 | Intron | XP_016880663.1 |