Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGCAACCCCCAAGCAGAACAGT[C/G]TCAGTCAGGCCACCCACCGTTTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611198 | ||||||||||||||||||||
Literature Links: |
JAKMIP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JAKMIP3 - Janus kinase and microtubule interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105521.3 | Intron | NP_001098991.1 | ||||
NM_001323086.1 | Intron | NP_001310015.1 | ||||
NM_001323087.1 | Intron | NP_001310016.1 | ||||
NM_001323088.1 | Intron | NP_001310017.1 | ||||
NM_001323089.1 | Intron | NP_001310018.1 | ||||
NM_001323090.1 | Intron | NP_001310019.1 | ||||
XM_005252674.4 | Intron | XP_005252731.1 | ||||
XM_005252676.4 | Intron | XP_005252733.1 | ||||
XM_011539676.2 | Intron | XP_011537978.1 | ||||
XM_011539682.2 | Intron | XP_011537984.1 | ||||
XM_017016095.1 | Intron | XP_016871584.1 | ||||
XM_017016096.1 | Intron | XP_016871585.1 | ||||
XM_017016097.1 | Intron | XP_016871586.1 | ||||
XM_017016098.1 | Intron | XP_016871587.1 | ||||
XM_017016099.1 | Intron | XP_016871588.1 |