Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGCACTGAAAAGCTTCCTCGTA[C/T]TGTCAAAGCCAGTAAGCCCCGAATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616745 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BOD1 - biorientation of chromosomes in cell division 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159651.1 | Intron | NP_001153123.1 | ||||
NM_138369.2 | Intron | NP_612378.1 | ||||
XM_017010017.1 | Intron | XP_016865506.1 |