Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603362 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3GL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SH3GL3 - SH3 domain containing GRB2 like 3, endophilin A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301108.1 | Intron | NP_001288037.1 | ||||
NM_001301109.1 | Intron | NP_001288038.1 | ||||
NM_001324182.1 | Intron | NP_001311111.1 | ||||
NM_001324183.1 | Intron | NP_001311112.1 | ||||
NM_001324184.1 | Intron | NP_001311113.1 | ||||
NM_001324185.1 | Intron | NP_001311114.1 | ||||
NM_001324186.1 | Intron | NP_001311115.1 | ||||
NM_001324187.1 | Intron | NP_001311116.1 | ||||
NM_003027.4 | Intron | NP_003018.3 | ||||
XM_011521889.1 | Intron | XP_011520191.1 | ||||
XM_011521891.2 | Intron | XP_011520193.1 | ||||
XM_011521892.2 | Intron | XP_011520194.1 | ||||
XM_017022486.1 | Intron | XP_016877975.1 |