Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTATGGATGGTTTTGAAGGACT[A/G]ATCCTCTTCTGCTGTCTGTGAACTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611505 MIM: 611534 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CENPP - centromere protein P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012267.2 | Intron | NP_001012267.1 | ||||
NM_001286969.1 | Intron | NP_001273898.1 | ||||
NM_001286971.1 | Intron | NP_001273900.1 | ||||
XM_011518685.2 | Intron | XP_011516987.1 | ||||
XM_011518689.1 | Intron | XP_011516991.1 | ||||
XM_017014715.1 | Intron | XP_016870204.1 |
NOL8 - nucleolar protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |