Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTCATTCTAAATGTCTTTATAG[G/T]CTATGAGAAATTATATACAAATAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PTPDC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPDC1 - protein tyrosine phosphatase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253829.1 | Intron | NP_001240758.1 | ||||
NM_001253830.1 | Intron | NP_001240759.1 | ||||
NM_152422.4 | Intron | NP_689635.3 | ||||
NM_177995.2 | Intron | NP_818931.1 | ||||
XM_005251711.4 | Intron | XP_005251768.1 | ||||
XM_011518240.2 | Intron | XP_011516542.1 | ||||
XM_011518241.2 | Intron | XP_011516543.1 | ||||
XM_017014280.1 | Intron | XP_016869769.1 | ||||
XM_017014281.1 | Intron | XP_016869770.1 | ||||
XM_017014282.1 | Intron | XP_016869771.1 |