Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGAGCTTCAGTCTGAGTCCCAGC[A/G]CCGAGAGCGTGTCTGACCGGTTGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609396 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHLPP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PHLPP1 - PH domain and leucine rich repeat protein phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194449.3 | 1311 | Missense Mutation | ACC,GCC | T,A 356 | NP_919431.2 | |
XM_011525886.1 | 1311 | Missense Mutation | ACC,GCC | T,A 356 | XP_011524188.1 |
Set Membership: |
HapMap |