Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTCATTAGCTCTGGGACTGTC[A/G]TACTGGGGGATAACCCTACTCTAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CCDC12 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC12 - coiled-coil domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277074.1 | 2527 | Intron | NP_001264003.1 | |||
NM_144716.5 | 2527 | Intron | NP_653317.2 | |||
XM_011533390.2 | 2527 | Intron | XP_011531692.1 | |||
XM_011533391.2 | 2527 | Intron | XP_011531693.1 | |||
XM_011533392.2 | 2527 | Intron | XP_011531694.1 | |||
XM_011533393.2 | 2527 | Intron | XP_011531695.1 | |||
XM_011533394.2 | 2527 | Intron | XP_011531696.1 | |||
XM_011533396.2 | 2527 | UTR 3 | XP_011531698.1 |