Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGTTCTTGGCTCCACAGCCCAT[G/T]GTGACATCACTTGGGGGCTCTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615144 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS55 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRSS55 - protease, serine 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197020.1 | 153 | Intron | NP_001183949.1 | |||
NM_198464.3 | 153 | Intron | NP_940866.2 | |||
XM_005272372.3 | 153 | Missense Mutation | ATG,ATT | M,I 31 | XP_005272429.2 | |
XM_011543814.2 | 153 | Missense Mutation | ATG,ATT | M,I 31 | XP_011542116.2 | |
XM_017013182.1 | 153 | Missense Mutation | ATG,ATT | M,I 31 | XP_016868671.1 |
Set Membership: |
HapMap |