Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGAAGATCACTTCAGCCCAGTC[A/G]TTCAAGTTCCAGCTTGGGCAACACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615435 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERO1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ERO1A - endoplasmic reticulum oxidoreductase 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014584.2 | 2907 | UTR 3 | NP_055399.1 | |||
XM_011536684.2 | 2907 | UTR 3 | XP_011534986.1 |
GPR137C - G protein-coupled receptor 137C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |