Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCGGCGTCCCGGGGGCGCAGGG[A/C]AGGAGGGCGTCCGAGTCAACAACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609217 MIM: 612326 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPIRE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPIRE2 - spire type actin nucleation factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCF25 - transcription factor 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014972.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | NP_055787.1 | |
XM_005256297.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_005256354.1 | |
XM_005256298.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_005256355.1 | |
XM_005256299.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_005256356.1 | |
XM_011522956.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_011521258.1 | |
XM_011522957.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_011521259.1 | |
XM_011522958.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_011521260.1 | |
XM_011522959.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_011521261.1 | |
XM_011522960.2 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_011521262.1 | |
XM_011522961.2 | 235 | UTR 5 | XP_011521263.1 | |||
XM_017023047.1 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_016878536.1 | |
XM_017023048.1 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_016878537.1 | |
XM_017023049.1 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_016878538.1 | |
XM_017023050.1 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_016878539.1 | |
XM_017023051.1 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_016878540.1 | |
XM_017023052.1 | 235 | Missense Mutation | AAG,CAG | K,Q 53 | XP_016878541.1 | |
XM_017023053.1 | 235 | Intron | XP_016878542.1 | |||
XM_017023054.1 | 235 | UTR 5 | XP_016878543.1 | |||
XM_017023055.1 | 235 | UTR 5 | XP_016878544.1 | |||
XM_017023056.1 | 235 | UTR 5 | XP_016878545.1 | |||
XM_017023057.1 | 235 | Intron | XP_016878546.1 | |||
XM_017023058.1 | 235 | Intron | XP_016878547.1 |