Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAGAGGAAGACTCAGCCAAGAC[A/T]GACAGAAGTATGACATGGCAAGGCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616167 MIM: 614709 | |||||||||||||||||||||||
Literature Links: |
DCUN1D3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DCUN1D3 - defective in cullin neddylation 1 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYRM1 - LYR motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128301.2 | Intron | NP_001121773.1 | ||||
NM_001128302.2 | Intron | NP_001121774.1 | ||||
NM_001302835.1 | Intron | NP_001289764.1 | ||||
NM_001302836.1 | Intron | NP_001289765.1 | ||||
NM_020424.4 | Intron | NP_065157.1 | ||||
XM_005255444.1 | Intron | XP_005255501.1 | ||||
XM_011545900.2 | Intron | XP_011544202.1 | ||||
XM_011545901.2 | Intron | XP_011544203.1 | ||||
XM_011545902.1 | Intron | XP_011544204.1 | ||||
XM_011545908.2 | Intron | XP_011544210.1 | ||||
XM_017023475.1 | Intron | XP_016878964.1 | ||||
XM_017023476.1 | Intron | XP_016878965.1 | ||||
XM_017023477.1 | Intron | XP_016878966.1 | ||||
XM_017023478.1 | Intron | XP_016878967.1 | ||||
XM_017023479.1 | Intron | XP_016878968.1 | ||||
XM_017023480.1 | Intron | XP_016878969.1 | ||||
XM_017023481.1 | Intron | XP_016878970.1 | ||||
XM_017023482.1 | Intron | XP_016878971.1 |