Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTCACAGCAACCTGGTGGGGTG[A/G]ATATGAACGTTCCCATTTTATACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616215 | ||||||||||||||||||||
Literature Links: |
CREB3L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CREB3L1 - cAMP responsive element binding protein 3 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052854.3 | Intron | NP_443086.1 | ||||
XM_006718380.3 | Intron | XP_006718443.1 |