Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATTATAATATTTGTATATACTG[G/T]ACTTAAGGACTTGAAAGATCACATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TTC17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTC17 - tetratricopeptide repeat domain 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307943.1 | Intron | NP_001294872.1 | ||||
NM_018259.5 | Intron | NP_060729.2 | ||||
XM_005253016.1 | Intron | XP_005253073.1 | ||||
XM_006718263.1 | Intron | XP_006718326.1 | ||||
XM_011520218.2 | Intron | XP_011518520.1 | ||||
XM_017018010.1 | Intron | XP_016873499.1 |