Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTTTTCTCAAGGTTGAAATTTCG[A/G]AAGTTCTGTTGGCAGCAGTTTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608817 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC4C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC4C - leucine rich repeat containing 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258419.1 | 7380 | Intron | NP_001245348.1 | |||
NM_020929.2 | 7380 | Intron | NP_065980.1 | |||
XM_011520238.2 | 7380 | UTR 3 | XP_011518540.1 | |||
XM_011520239.2 | 7380 | UTR 3 | XP_011518541.1 | |||
XM_011520240.2 | 7380 | UTR 3 | XP_011518542.1 | |||
XM_011520241.2 | 7380 | UTR 3 | XP_011518543.1 | |||
XM_011520242.2 | 7380 | UTR 3 | XP_011518544.1 | |||
XM_011520243.2 | 7380 | UTR 3 | XP_011518545.1 | |||
XM_011520244.2 | 7380 | UTR 3 | XP_011518546.1 | |||
XM_017018070.1 | 7380 | UTR 3 | XP_016873559.1 | |||
XM_017018071.1 | 7380 | UTR 3 | XP_016873560.1 | |||
XM_017018072.1 | 7380 | UTR 3 | XP_016873561.1 | |||
XM_017018073.1 | 7380 | UTR 3 | XP_016873562.1 | |||
XM_017018074.1 | 7380 | UTR 3 | XP_016873563.1 | |||
XM_017018075.1 | 7380 | UTR 3 | XP_016873564.1 | |||
XM_017018076.1 | 7380 | UTR 3 | XP_016873565.1 | |||
XM_017018077.1 | 7380 | UTR 3 | XP_016873566.1 | |||
XM_017018078.1 | 7380 | UTR 3 | XP_016873567.1 | |||
XM_017018079.1 | 7380 | UTR 3 | XP_016873568.1 |