Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGACTGTTCTGTGAACTCATGA[C/T]AGTGCATTTCAAGAAATGTAGTGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179616 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf74 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C11orf74 - chromosome 11 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276722.1 | Intron | NP_001263651.1 | ||||
NM_001276723.1 | Intron | NP_001263652.1 | ||||
NM_001276724.1 | Intron | NP_001263653.1 | ||||
NM_001276725.1 | Intron | NP_001263654.1 | ||||
NM_001276726.1 | Intron | NP_001263655.1 | ||||
NM_001276727.1 | Intron | NP_001263656.1 | ||||
NM_138787.3 | Intron | NP_620142.2 | ||||
XM_006718138.3 | Intron | XP_006718201.1 | ||||
XM_011519885.2 | Intron | XP_011518187.1 |
RAG2 - recombination activating gene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |