Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAATTTTACCCATTTTAAGGAA[C/T]ATAGTCCAATATAATAATCCACGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609154 | ||||||||||||||||||||
Literature Links: |
ASCL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASCL3 - achaete-scute family bHLH transcription factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM9B - TMEM9 domain family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286094.1 | Intron | NP_001273023.1 | ||||
NM_001286095.1 | Intron | NP_001273024.1 | ||||
NM_020644.2 | Intron | NP_065695.1 |