Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAAATTACTTTCTGGATTTCAAC[A/G]TTTAAGGCTGATTTCTTCTCAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611578 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRRS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRRS1 - ferric chelate reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013660.2 | 1660 | Intron | NP_001013682.2 | |||
XM_005270861.3 | 1660 | Intron | XP_005270918.1 | |||
XM_011541451.2 | 1660 | Intron | XP_011539753.1 | |||
XM_011541452.1 | 1660 | UTR 3 | XP_011539754.1 | |||
XM_017001271.1 | 1660 | Intron | XP_016856760.1 |
Set Membership: |
HapMap |