Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCCATTTGGAGGGTGGCAGCT[C/T]AGGCTCAGCCCCCAGGACACTCCCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607582 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC22A6 - solute carrier family 22 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004790.4 | Intron | NP_004781.2 | ||||
NM_153276.2 | Intron | NP_695008.1 | ||||
NM_153277.2 | Intron | NP_695009.1 | ||||
NM_153278.2 | Intron | NP_695010.1 | ||||
XM_017018562.1 | Intron | XP_016874051.1 |
Set Membership: |
HapMap |