Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAATAAAGTTCTGTTCTTATGCA[G/T]AACTCTCAAAAACAAAACCCCTCAA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 609369 | |||||||||||||||||||||||||||||
Literature Links: |
ATL3 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ATL3 - atlastin GTPase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290048.1 | 3158 | UTR 3 | NP_001276977.1 | |||
NM_015459.4 | 3158 | UTR 3 | NP_056274.3 | |||
XM_006718493.1 | 3158 | UTR 3 | XP_006718556.1 |
Set Membership: |
HapMap |