Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGCAGAAACTGACTCGTGCCCT[A/C]TATTGATATAAAATCCCCCAGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAA40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NAA40 - N(alpha)-acetyltransferase 40, NatD catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300800.1 | Intron | NP_001287729.1 | ||||
NM_024771.3 | Intron | NP_079047.2 | ||||
XM_006718689.3 | Intron | XP_006718752.1 | ||||
XM_011545254.2 | Intron | XP_011543556.1 | ||||
XM_011545255.2 | Intron | XP_011543557.1 |
Set Membership: |
HapMap |