Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGGGAAAATGAAGCCCAACAT[C/T]TCCACTGTGGCCAATAAGGCCATGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602905 MIM: 605161 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
KCNS1 - potassium voltage-gated channel modifier subfamily S member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WFDC5 - WAP four-disulfide core domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145652.3 | Intron | NP_663627.1 | ||||
XM_011528601.1 | Intron | XP_011526903.1 | ||||
XM_011528602.1 | Intron | XP_011526904.1 |
Set Membership: |
HapMap |