Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATAGGGTTTTTCTCCTGTGTGA[A/G]TTCTCTGATGTCTTTCAACTGAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf229 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf229 - chromosome 1 open reading frame 229 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF669 - zinc finger protein 669 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142572.1 | 694 | Missense Mutation | ACT,ATT | T,I 185 | NP_001136044.1 | |
NM_024804.2 | 694 | Missense Mutation | ACT,ATT | T,I 271 | NP_079080.2 | |
XM_005273270.2 | 694 | Missense Mutation | ACT,ATT | T,I 148 | XP_005273327.1 | |
XM_005273271.2 | 694 | Missense Mutation | ACT,ATT | T,I 148 | XP_005273328.1 |