Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATGTAAAATTGTATTTACCAT[A/G]TAAGTTCAGCTTTAAAAACAATAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607365 | |||||||||||||||||||||||
Literature Links: |
LIPH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LIPH - lipase H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139248.2 | Intron | NP_640341.1 | ||||
XM_006713529.3 | Intron | XP_006713592.1 | ||||
XM_011512530.2 | Intron | XP_011510832.1 | ||||
XM_011512531.2 | Intron | XP_011510833.1 | ||||
XM_017005852.1 | Intron | XP_016861341.1 |