Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMC8 - armadillo repeat containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267041.2 | Intron | NP_001253970.1 | ||||
NM_001267042.2 | Intron | NP_001253971.1 | ||||
NM_001282342.1 | Intron | NP_001269271.1 | ||||
NM_014154.4 | Intron | NP_054873.2 | ||||
NM_015396.5 | Intron | NP_056211.2 | ||||
NM_213654.3 | Intron | NP_998819.1 | ||||
XM_006713564.3 | Intron | XP_006713627.1 | ||||
XM_006713565.2 | Intron | XP_006713628.1 | ||||
XM_006713566.3 | Intron | XP_006713629.1 | ||||
XM_006713567.3 | Intron | XP_006713630.1 | ||||
XM_011512632.1 | Intron | XP_011510934.1 | ||||
XM_011512633.1 | Intron | XP_011510935.1 | ||||
XM_017006095.1 | Intron | XP_016861584.1 | ||||
XM_017006096.1 | Intron | XP_016861585.1 | ||||
XM_017006097.1 | Intron | XP_016861586.1 | ||||
XM_017006098.1 | Intron | XP_016861587.1 | ||||
XM_017006099.1 | Intron | XP_016861588.1 | ||||
XM_017006100.1 | Intron | XP_016861589.1 | ||||
XM_017006101.1 | Intron | XP_016861590.1 |