Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCATGTATTTGCCTCTCCCTCCC[C/T]TTAACTCCTAGCAATCACTGATCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C14orf132 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf132 - chromosome 14 open reading frame 132 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252507.2 | Intron | NP_001239436.1 | ||||
NM_001282463.1 | Intron | NP_001269392.1 | ||||
NM_001282464.1 | Intron | NP_001269393.1 | ||||
NM_001289139.1 | Intron | NP_001276068.1 | ||||
XM_017021469.1 | Intron | XP_016876958.1 |