Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGGTTCTTTTTTAAGTTTCTCG[G/T]TCTTTTTTTTTTTTTTTTTTTTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194648 | ||||||||||||||||||||
Literature Links: |
MIR571 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR571 - microRNA 571 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF141 - zinc finger protein 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003441.2 | Intron | NP_003432.1 | ||||
XM_011513562.2 | Intron | XP_011511864.1 | ||||
XM_011513563.2 | Intron | XP_011511865.1 | ||||
XM_017008591.1 | Intron | XP_016864080.1 |