Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGAAACTGGTCAGCAGACTCC[A/G]GCTACATAGGTTATGAATTCTCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604741 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR1D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKR1D1 - aldo-keto reductase family 1, member D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190906.1 | Intron | NP_001177835.1 | ||||
NM_001190907.1 | Intron | NP_001177836.1 | ||||
NM_005989.3 | Intron | NP_005980.1 |
Set Membership: |
HapMap |