Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCACATGCTTTCACATGTGAATA[C/T]AGCAGGAATGAAAAAAGAGCATAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603972 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF43 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF43 - zinc finger protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256648.1 | 9276 | Intron | NP_001243577.1 | |||
NM_001256649.1 | 9276 | UTR 3 | NP_001243578.1 | |||
NM_001256650.1 | 9276 | Intron | NP_001243579.1 | |||
NM_001256651.1 | 9276 | Intron | NP_001243580.1 | |||
NM_001256653.1 | 9276 | Intron | NP_001243582.1 | |||
NM_001256654.1 | 9276 | UTR 3 | NP_001243583.1 | |||
NM_003423.3 | 9276 | UTR 3 | NP_003414.2 | |||
XM_011528257.2 | 9276 | Intron | XP_011526559.1 | |||
XM_011528259.2 | 9276 | UTR 3 | XP_011526561.1 | |||
XM_017027207.1 | 9276 | Intron | XP_016882696.1 | |||
XM_017027208.1 | 9276 | Intron | XP_016882697.1 | |||
XM_017027209.1 | 9276 | Intron | XP_016882698.1 | |||
XM_017027210.1 | 9276 | Intron | XP_016882699.1 | |||
XM_017027211.1 | 9276 | UTR 3 | XP_016882700.1 | |||
XM_017027212.1 | 9276 | Intron | XP_016882701.1 | |||
XM_017027213.1 | 9276 | Intron | XP_016882702.1 | |||
XM_017027214.1 | 9276 | Intron | XP_016882703.1 | |||
XM_017027215.1 | 9276 | Intron | XP_016882704.1 | |||
XM_017027216.1 | 9276 | Intron | XP_016882705.1 |
Set Membership: |
HapMap |