Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAGGAAGACGCTGGTTTACATA[A/T]AGCAGAATTTGGAAATATTCTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603795 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LITAF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LITAF - lipopolysaccharide induced TNF factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136472.1 | Intron | NP_001129944.1 | ||||
NM_001136473.1 | Intron | NP_001129945.1 | ||||
NM_004862.3 | Intron | NP_004853.2 | ||||
XM_006720982.2 | Intron | XP_006721045.1 | ||||
XM_006720983.3 | Intron | XP_006721046.1 | ||||
XM_006720984.3 | Intron | XP_006721047.1 | ||||
XM_006720985.3 | Intron | XP_006721048.1 | ||||
XM_011522754.2 | Intron | XP_011521056.1 | ||||
XM_017023896.1 | Intron | XP_016879385.1 |
Set Membership: |
HapMap |