Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTAGCTCTGTCATTTGCAGTGCC[A/G]CCTTAGGCAATCGACTCAGCTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608989 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDR16C5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SDR16C5 - short chain dehydrogenase/reductase family 16C, member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318049.1 | Intron | NP_001304978.1 | ||||
NM_001318050.1 | Intron | NP_001304979.1 | ||||
NM_138969.3 | Intron | NP_620419.2 | ||||
XM_011517479.1 | Intron | XP_011515781.1 |