Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607216 | ||||||||||||||||||||
Literature Links: |
SDK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDK1 - sidekick cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079653.1 | Intron | NP_001073121.1 | ||||
NM_152744.3 | Intron | NP_689957.3 | ||||
XM_011515188.1 | Intron | XP_011513490.1 | ||||
XM_011515189.1 | Intron | XP_011513491.1 | ||||
XM_011515190.1 | Intron | XP_011513492.1 | ||||
XM_017011837.1 | Intron | XP_016867326.1 | ||||
XM_017011838.1 | Intron | XP_016867327.1 | ||||
XM_017011839.1 | Intron | XP_016867328.1 | ||||
XM_017011840.1 | Intron | XP_016867329.1 |