Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCGCCTCATCTCCCGGAATGT[A/T]TCAAAGGAAACCCCTGTCTTTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609964 | ||||||||||||||||||||||||||||||||
Literature Links: |
CLEC4D PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CLEC4D - C-type lectin domain family 4 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080387.4 | 31 | Intron | NP_525126.2 | |||
XM_011520632.2 | 31 | UTR 5 | XP_011518934.1 | |||
XM_011520633.2 | 31 | Intron | XP_011518935.1 |
Set Membership: |
HapMap |