Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACTACATGATCTCACTTGTTTG[A/T]GGAATCCAAAATAGTCAAACTCATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CXorf56 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CXorf56 - chromosome X open reading frame 56 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170569.1 | Intron | NP_001164040.1 | ||||
NM_001170570.1 | Intron | NP_001164041.1 | ||||
NM_022101.3 | Intron | NP_071384.1 |